Would you like to log in again?
VWR MAT PURESTEP BLUE 46X60 CASE OF 4
Supplier: VWR International
VWR* Purestep Mat, 46X60 30 Layer Blue^ VWR* Purestep Mat, 46X60 30 Layer Blue, Removes/Contains Dirt And Dust From Foot Traffic And Equipment Wheels.Consists Of 30 Sheets Of 1.5Mil Ldpe Film.Each Sheet Has A 0.3Mil Thick Acrylic-Based, Pressure-Sensitive Adhesive Coating With Antimicrobial Agent
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR MAT PURESTEP BLUE 46X60 CASE OF 4 76196-346 Case of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty