Would you like to log in again?
Endoglycoceramidase I (EGCase I)
Supplier: New England Biolabs
P0773S
76196-404EA 83 USD InStock
76196-404
Endoglycoceramidase I (EGCase I)
Enzymes
Endoglycoceramidase I (EGCase I) catalyzes the hydrolysis of the β-glycosidic linkage between oligosaccharides and ceramides in various glycosphingolipids.
  • Cleaves the β-glycosidic linkage between oligosaccharides and ceramides in glycosphingolipids
  • Oligosaccharide and ceramide remain intact, leaving them suitable for further analysis
  • This is an Enzyme for Innovation (EFI), a project initiated by NEB to provide unique enzymes
The enzyme leaves the oligosaccharide and ceramide intact, keeping them suitable for further analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty