Would you like to log in again?
Recombinant 1-2,4,6 Fucosidase O
Supplier: New England Biolabs
P0749S P0749L
76196-418EA 151 USD InStock
76196-418 76196-416
Recombinant 1-2,4,6 Fucosidase O
Enzymes
a1-2,4,6 Fucosidase O is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal a1-2, a1-4 and a1-6 linked fucose residues from oligosaccharides. a1-2,4,6 Fucosidase O cleaves a1-6 fucose residues more efficiently than other linkages.
  • Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity
  • Glycerol -free for optimal performance in HPLC and mass spectrometry analysis
  • ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Optimal activity and stability for up to 12 months
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty