Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Top Dog 6 Mil Die Cut Apron, 50" Length, Remco Products

Supplier: Remco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Top Dog
80026 80024 80025 80023 80022
76196-566CS 617.67 USD InStock
76196-566 76196-570 76196-516 76196-514 76196-568
Top Dog 6 Mil Die Cut Apron, 50" Length, Remco Products
Workwear
Reusable and washable, the one-piece construction increases the apron's hygiene and durability.

  • Resistant to chemical and acid sanitation products
  • Non-porous surface will not host bacteria or mold
  • Washer and dryer safe
  • Will not crack or stiffen in low temperatures or with repeated cleaning
  • Impervious to animal fats and degreasers
  • Completely latex-free for sensitive users
  • Easily repairable

6 mil aprons from Top Dog come in five colors to support color-coded HACCP plans and zoning efforts. The 6 Mil Die Cut Apron is made of pure polyurethane and is reversible. It fits comfortably over clothes. One-piece construction increases the apron's hygiene and durability. The apron is machine washable, dryer-safe to 180 °F, and chemical and abrasive resistant. The 50" length makes the apron cover most of an employee's torso and legs without becoming cumbersome.

Certifications: Made of FDA Compliant material.

Ordering information: Aprons can only be purchased by the case. Case quantity is 36.

Delivery information: Dropshipping available.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty