Would you like to log in again?
CARTRIDGE FP SELECT SILICA 80G BX10
Supplier: Buchi
FlashPure Select Silica catridge 80g^ FlashPure Select Silica catridge, Particle shape: Spherica, Particle size: 13-17 um, Pore size: 30-70A, Surface area: 600-800m2/g, Max.Pressure: 350psi / 24bar, Default flowrate: 60 mL/min, Loading capacity: 80mg-24g, Column volume: 120 mL, weight: 80g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE FP SELECT SILICA 80G BX10 76196-932 140000117 Box of 10 (80g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty