Would you like to log in again?
ADAPTOR FP EMPTY SOLID LOADER 28 FITTING
Supplier: Buchi
FlashPure Empty Solid Loader Adaptor Kit^ FlashPure Empty Solid Loader Adaptor Kit, incl. SS male luer lock to 1/4"-28 fitting, 1/4-28 to 5/16In-24 PEEK adapter and o-rings.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTOR FP EMPTY SOLID LOADER 28 FITTING 76197-008 145173491 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty