Would you like to log in again?
FlashPure EcoFlex Cartridges, Büchi
Supplier: Büchi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
140000078 140000039 140000030 140000056 140000100 140000095 140000076 140000080 140000051 140000081 140000026 140000040 140000103 140000065 140000036 140000104 140000050 140000027 140000058 140000035 140000096 140000037 140000083 140000046 140000069 140000041 140000054 140000064 140000085 140000052 140000066 140000082 140000024 140000060 140000047 140000075 140000061 140000031 140000102 140000091 140000032 140000071 140000045 140000055 140000059 140000044 140000077 140000087 140000098 140000097 140000043 140000063 140000099 140000062 140000022 140000072 140000067 140000093 140000023 140000094 140000029 140000028 140000073 140000053 140000089 140000049 140000033 140000034 140000079 140000092 140000048 140000070 140000090 140000068 140000038 140000057 140000025 140000084 140000088 140000042 140000101 140000074 140000086 140000021
76196-874BX 247.79 USD InStock
76196-874 76196-830 76196-796 76196-872 76196-794 76196-870 76196-792 76196-790 76196-838 76196-836 76197-040 76196-834 76196-878 76196-876 76196-832 76196-798 76197-046 76197-048 76197-042 76197-044 76196-828 76196-840 76196-884 76196-882 76196-880 76196-848 76196-804 76196-846 76196-802 76196-844 76196-800 76196-888 76196-842 76196-886 76197-034 76197-036 76197-030 76197-032 76197-038 76196-852 76196-850 76196-892 76196-890 76197-060 76196-816 76196-814 76197-062 76196-858 76196-812 76196-856 76196-854 76196-810 76197-024 76197-026 76197-020 76196-808 76197-022 76196-806 76197-028 76196-786 76196-862 76196-784 76196-860 76196-782 76196-780 76196-826 76197-050 76196-868 76196-824 76196-822 76196-866 76196-788 76196-864 76196-820 76197-056 76197-012 76197-058 76197-014 76197-052 76196-818 76197-054 76197-010 76197-016 76197-018
FlashPure EcoFlex Cartridges, Büchi
Chromatography Columns
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries. This enables the user to choose the flash cartridge which best suits his purification needs.
  • Different sizes for scalability from milligrams to grams
  • Broad range of different normal and reversed phases
  • Small and spherical particles for fast separations and high loadings
Compatibility with the Pure Navigator software (flash method optimization by interpreting TLC or HPLC scouting runs). RFID tag for an automatic transfer of important parameters for a run on a Pure chromatography system.
Silica phase are high and medium polar compounds, most popular polar phase, mainly used for flash applications. C18 WP (wide pores) has a ideal application in peptides.
C18 phase are low polar and non-polar compounds, most popular non-polar phase. Amino phase are high and medium polar molecules example carbohydrates and amines. Alumina Wide variety of molecules example acidic, basic or acid sensitive compounds.
Diol, C18 and Amino phase has spherical particle shape. Silica and alumina has irregular shape.
Loading capacity: 0.1 to 10% for Silica, Silica LOK, Silica Solid Loader; 0.1 to 4% for Alumina Neutra, Alumina Basic, Alumina Acidic; 0.1 to 2.5% for C18; 0.1 to 5% for Amino, Diol.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty