Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Reagent Troughs
Supplier: TradeWinds (TWD)
CT-L-100 CT-RG-100
76197-168CS 584.99 USD InStock
76197-168 76197-170
Reagent Troughs
Reagent Reservoirs
Reagent Troughs are made of polypropylene. Troughs support a disposable polypropylene trough liner. Liners are made of thin, disposable, lightweight polypropylene.
  • V-shaped bottom and angled sides for maximum reagent recovery
  • Usable with up to 12-channel pipettors
  • Designed for SBS format applications and compatible with robotic systems
  • Reusable
Perfect for 12 or 8-channel pipettes and compatible eith robotic systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty