Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-His Tag Rabbit Monoclonal Antibody [clone: 25B6E11]

Supplier: Genscript
GenScript®
A01857-40
76197-310EA 269.34 USD InStock
76197-310
Anti-His Tag Rabbit Monoclonal Antibody [clone: 25B6E11]
Antibodies
GenScript MonoRab™ anti-His tag (C-term) antibody (25B6E11), mAb, rabbit specifically recognizes His tags placed at C-terminal and internal regions of His-fusion
proteins. This product is not recommended for N-terminal His-fusion protein detection and nbsp.

  • Comprehensive pool
  • against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

The lyophilized product remains stable for up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 to 3 weeks at 2 to 8 °C or for up to 12 months at −20 °C or below. Upon receiving, spin the vial down to place the lyophilized antibody on the bottom of the vial. Reconstitute the antibody as mentioned above. Prepare single use aliquots (no less than 10 µl) in amber vials and freeze it immediately at −20 °C. The frozen aliquots should be thawed once with any remainder kept at 4 °C.

Type: Tag
Antigen: His Tag
Clonality: Monoclonal
Clone: 25B6E11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty