Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pembrolizumab Mouse Monoclonal Antibody [clone: 1D6E10]

Supplier: GenScript Biotech
GenScript®
A01844-40
76198-386EA 189.21 USD InStock
76198-386
Anti-Pembrolizumab Mouse Monoclonal Antibody [clone: 1D6E10]
Antibodies
The product is specific for Pembrolizumab.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Pembrolizumab
Clonality: Monoclonal
Clone: 1D6E10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty