Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PEG Mouse Monoclonal Antibody [clone: 5E10E9]

Supplier: GenScript Biotech
GenScript®
A01795-100 A01795-500
76198-876EA 205.68 USD InStock
76198-876 76198-878
Anti-PEG Mouse Monoclonal Antibody [clone: 5E10E9]
Antibodies
GenScript’s THE™ PEG Antibody, mAb, Mouse has high affinity for the PEG backbone. The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates. and nbsp.

  • Anti-Drug antibodies
  • High specificit
  • High affinity
  • High stability

GenScript’s THE™ PEG Antibody, mAb, Mouse has high affinity for the PEG backbone. The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates.

Type: Primary
Antigen: PEG
Clonality: Monoclonal
Clone: 5E10E9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty