Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PD-1 Mouse Monoclonal Antibody [clone: PD1.D3]

Supplier: Genscript
GenScript®
A01829-40
76198-898EA 148.24 USD InStock
76198-898
Anti-PD-1 Mouse Monoclonal Antibody [clone: PD1.D3]
Antibodies
The product is specific for human PD-1 (UniProt Accession: Q15116). This antibody blocks human PD-L1 binding with human PD-1 expression cell line in flow cytometry assay.

  • Immune checkpoint antibodies
  • Positive control for antibody drug early development
  • Validate cell line development
  • Facilitate assay development

To facilitate this hot immunotherapy research field, GenScript provides immune checkpoint antibodies and immune checkpoint proteins for many kinds of applications. Products for more targets are coming in succession.
.

Type: Primary
Antigen: PD-1
Clonality: Monoclonal
Clone: PD1.D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty