Would you like to log in again?
eStain™ L1 Protein Staining Device, GenScript
Supplier: GenScript Biotech
eStain™
L00660 L00662-10 L00661-5 L00661-10 L00663 L00753 L00657 L00658
76198-948EA 50.15 USD InStock
76198-948 76510-402 76198-950 76198-946 76510-486 76510-398 76198-944 76198-954
eStain™ L1 Protein Staining Device, GenScript
Staining Boxes
The eStain™ is a fully automated protein PAGE gel staining system, which uses Coomassie Brilliant Blue and a patented protein staining technology. The system integrates the traditional three steps of fixing, staining, and destaining into one automated process. It can stain and destain two protein PAGE gels simultaneously in 10 minutes.
  • Automated 10-minute staining and destaining for 2 gels simultaneously
  • Consistent results with coomassie blue staining (offered by GenScript) or membrane staining with Ponceau S solution
  • Easy to use with clean setup and convenient operation
  • Sharply stained protein bands with sensitivity ≥12.5 ng
The eStain™ system works with all types of precast as well as homemade PAGE gels. Compared with the conventional staining methods, gels stained by eStain™ have crisp blue bands with minimum or no background. The eStain™ also offers superior sensitivity compared to conventional methods and can detect as low as 12.5ng of protein. The shortened workflow increases productivity. This protein staining system combines three steps of traditional Coomassie blue staining method into one single step to greatly reduce the time required for protein staining.
Excellent uniformity and repeatability with either Coomasie Blue and Ponceau S solution.
Ordering information: eStain L1C Staining Kit (Cat No: L00753) contains reagents that are used with eStain L1 protein staining device (L00657). Each kit contains 1 x 2L of staining solution, 1 x 2L of destaining solution, and a pack of filter paper (50 pK).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty