Would you like to log in again?
PLATE LOW BASE SCREENSTAR ST PS BLK CS68
Supplier: Greiner Bio-One
Ultra Low Base SCREENSTAR Plate 1536 well^ULtra Low Base SCREENSTAR Plate,CO, material: Polystyrene, Sterile, Cell-Repellent surface, flat Bottom, color: Black uClear, with Lid, Spheroid culture of tumor cells, for suspension and spheroid culture, 4 years shelf life
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE LOW BASE SCREENSTAR ST PS BLK CS68 76199-184 789979 Case of 68 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty