Would you like to log in again?
TUBE CRYO.S W/96 YELLOW-CAP 300UL CS960
Supplier: Greiner Bio-One
Cryo.s* tube with 96 yellow-capped 300ul^ Cryo.s* tube with 96 yellow-capped, pre-racked, Datamatrix Coding Service for Cryogenic Storage Tubes, Ultra-sharp 2D code, Zero leachate polypropylene, medical grade and USP class VI certified, Medical grade silicone1 gasket, size: 300 ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE CRYO.S W/96 YELLOW-CAP 300UL CS960 76199-192 976585 Case of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty