Would you like to log in again?
STORAGE BOX 96VESSEL PC 1.2 ML CASE 1
Supplier: Greiner Bio-One
Storage box 96 vessel^ Storage box, Material: polycarbonate, Comes with a coding card, which enables proper storage of samples, capacity: 1.2 ml each, temperature: -80deg C to +121deg C, All components are autoclavable, size: 96 vessel
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STORAGE BOX 96VESSEL PC 1.2 ML CASE 1 76199-194 975502 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty