Would you like to log in again?
PLATE STRIP MICROLON 200 12XC8 WL CS150
Supplier: Greiner Bio-One
Microlon*200 Single-break Strip Plate 96 well^ Microlon*200, Single-break Strip Plate, Medium Binding, Material: polystyrene, color: clear, 1x8 Break, Well profile: C-bottom (flat bottom profile with curved edge Strips), Free of detectable DNase, RNase, human DNA, Non-Pyrogenic, size: 96 well
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE STRIP MICROLON 200 12XC8 WL CS150 76199-196 705070-25 Case of 150 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty