Would you like to log in again?
MICROPLATE 384 WL STERILE W/LID PS CS32
Supplier: Greiner Bio-One
MicroPlate 384 well^ MicroPlate, material: Polystyrene, Sterile, Cell-Repellent surface, Round (U) Bottom, color: Clear, with Lid, Spheroid culture of tumor cells, Spheroid culture of embryonic cells, Embryoid body formation, for suspension and spheroid culture, 4 years shelf life, 384 well
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROPLATE 384 WL STERILE W/LID PS CS32 76199-198 787979 Case of 32 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty