Would you like to log in again?
Protein A Agarose (Endotoxin-free), BioVision, Inc.
Supplier: BioVision
AB286896-1ML AB286896-25ML AB286896-5ML
76199-240EA 1468.81 USD InStock
76199-240 76199-232 76199-238
Protein A Agarose (Endotoxin-free), BioVision, Inc.
Electrophoresis Gels Agarose and Acrylamide
Reusable, Endotoxin free Protein A conjugated Agarose beads with ≥15 mg/ml binding capacity
  • APPLICATIONS
  • Purification of monoclonal and polyclonal antibodies from culture media, serum, ascites fluid or hybridoma supernatants.
  • Isolation of antibody/antigen complexes in immunoprecipitation experiments, since only the Fc region is involved in antibody binding and the Fab region is available for binding antigen.
Endo toxin free Protein A-Agarose beads are prepared by covalently coupling recombinant Protein A to 6% cross-linked Agarose beads under proprietary endotoxin-free conditions. BioVision’s Protein A (Cat. No. 6500, Cat. No. 6500B) is a genetically engineered protein containing five IgG-binding regions of native Protein A. The cell wall binding region, albumin binding region and other non-specific regions have been eliminated from the recombinant Protein A to ensure the maximum specific IgG binding. Protein A-Agarose beads display high chemical & physical stability as well as high flow rate, hydrophilicity & high gel strength. It can be used for IgG purification and immunoprecipitation. Protein A-Agarose beads are prepared by covalently coupling recombinant Protein A to 6% cross-linked Agarose beads. The coupling technique is optimized to give a higher binding capacity for IgG & minimum leaching of recombinant Protein A. The IgG binding capacity of Protein A-Agarose is ≥ 15 mg human or rabbit IgG per ml of settled beads. Our Endotoxin-free Protein A-Agarose also shows ability of reducing/removing certain amount of endotoxin from serum or ascites samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty