Would you like to log in again?
BioSim™ anti-Pembrolizumab (Keytruda®) (Human) ELISA Kit, BioVision
Supplier: BioVision
BioSim™
AB237666-96
76199-678EA 1691.87 USD InStock
76199-678
BioSim™ anti-Pembrolizumab (Keytruda®) (Human) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit for the qualitative determination of antibody against Pembrolizumab.
  • Easy, convenient and time-saving method
Pembrolizumab (Keytruda®) is an antibody drug that targets the cell surface receptor programmed cell death protein 1 (PD-1) found on T cells. By preventing the binding of its ligands (PD-L1 and PD-L2), pembrolizumab induces an antitumor immune response. Upregulation of PD-1 ligands is a mechanism for tumors to evade antitumor immune response; when PD-1 binds its ligand, the T cell receives an inhibitory signal leading to T cell anergy and blockade of anti-tumour immune response. Instead of directly targeting tumor tissue to induce tumor cell death, pembrolizumab acts as a checkpoint inhibitor to stimulate immune responses to eliminate cancer cells. However, some patients develop unwanted immunogenicity, which leads to production of anti-drug-antibodies (ADAs) inactivating the therapeutic effects of the treatment and, in rare cases, inducing adverse effects.
BioVision's BioSimTM anti-Pembrolizumab ELISA kit is designed to detect antibody against Pembrolizumab with high specificity and sensitivity in biological matrices. Blood sampling time is critical for detection of ATP. It is convenient to obtain blood sample just before the infusion or at least 2 weeks after the infusion of Pembrolizumab.
Cross Reactivity: Pembrolizumab (Keytruda®) infusion camouflages/masks the presence of antibody to Pembrolizumab (ATP) in serum/plasma samples. Therefore, blood sampling time is critical for detection of ATP. It is convenient to obtain blood sample just before the infusion or at least 2 weeks after the infusion of Pembrolizumab.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty