Would you like to log in again?
EZClick™ Global Protein Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision, Inc.
Supplier: BioVision
AB235634-100TEST
76200-046EA 880.68 USD InStock
76200-046
EZClick™ Global Protein Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision, Inc.
Assays Protein Assays
Assay to screen/study/characterize thioredoxin 1 inhibitors.
  • Rapid, simple and sensitive
  • Convenient and HTS
  • Includes inhibitor control
The human Thioredoxin 1 (Trx1) is a key enzyme that protects cells from oxidative stress by reducing disulfides and methionine sulfoxide groups in oxidized proteins. It also regulates DNA repair and apoptosis. In mammals, Trx1 is necessary for growth and development. However, an aberrant elevated expression of Trx1 is known to promote lung, cervical, pancreatic, colorectal, and breast cancers.
The screening kit, Trx1 reduces quenched di-FITC-labeled oxidized glutathione (Trx1 substrate), to mono-FITC-labeled unquenched reduced glutathione. In the presence of a Trx1 inhibitor (control inhibitor included), the reaction is impeded resulting in lower or no signal. The assay is high-throughput adaptable and can be completed in less than 1 hr.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty