Would you like to log in again?
Soluble Epoxide Hydrolase Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB283390-100TEST
76200-052EA 1147.12 USD InStock
76200-052
Soluble Epoxide Hydrolase Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simplest high-throughput assay to screen thioredoxin 1 inhibitors.
  • Rapid, simple and sensitive
  • Convenient and HTS
  • With NCND (inhibitor control)
Soluble Epoxide Hydrolase (EC 3.3.2.10) is a cytosolic enzyme with highest expression in the liver but present ubiquitously in several organs including the kidney, pancreatic islets, pituitary gland, lymphoid tissues, muscles, and the gastrointestinal tract. It catalyzes the hydrolysis of epoxyeicosatrienoic acids (EETs), i.e., epoxides derived from cytochrome P-450 mediated metabolism of arachidonic acid, to form vicinal diols.
The EETs cause vasodilation and have anti-inflammatory actions. Since the EETs are hydrolyzed to diols through soluble epoxide hydrolase, the inhibition of this enzyme is likely to enhance the beneficial properties of EETs. This makes soluble epoxide hydrolase a very useful target for cardiovascular and pain medicine.
This kit is a micro-plate based fluorometric kit for screening sEH inhibitors. Its assay principle is based on the hydrolysis of a non-fluorescent substrate to a fluorescent product by sEH. It uses N-Cyclohexyl-Nʹ-dodecylurea (NCND), a well-known sEH inhibitor, as positive control. The kit provides a simple and fast method to screen/ characterize potential sEH inhibitors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty