Would you like to log in again?
PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB235670-100TEST
76200-060EA 874.67 USD InStock
76200-060
PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric), BioVision
Assays Cellular Assays
It specifically detects reduced glutathione. Oxidized GSSG is not detected.
  • Simple procedure, takes only approximately 1 to 2 hours
  • Fast and convenient
  • Specific: does not detect oxidized GSSG
  • Sensitive: detects as low as 8 pmol GSH
Glutathione (GSH), a thiol-containing tripeptide, is a key antioxidant in living systems. Intracellular GSH status appears to be a sensitive indicator of the overall health of a cell. THe kit is based on a specific enzymatic cycling method in the presence of GSH and a fluorophore.
The reduction of the fluorophore produces a stable fluorescent product, the fluorescence of which is directly proportional to the amount of GSH in the sample and can be followed kinetically (Ex/Em = 535/587 nm). Our assay is the most specific and sensitive in the market. Oxidized Glutathione does not interfere with the assay. The assay is simple, reproducible and can specifically detect as low as 8 pmol/well of reduced form of Glutathione (GSH) in a 100 µl reaction
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty