Would you like to log in again?
Corning® LSE™ Multiposition Digital Vortexer, 230 V
Supplier: Corning
Corning®
6710
76200-270EA 1486.1 USD InStock
76200-270
Corning® LSE™ Multiposition Digital Vortexer, 230 V
Shakers and Mixers Vortex Mixers
The Corning® LSE multipostion digital vortexer is a multipurpose shaker dedicated to a variety of aggressive shaking and mixing applications.
  • 230 V model
  • Compact design with easy-access control panel
  • Safe for cold room or incubator use
  • Five-year warranty
Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform features spring-loaded retaining bars that adjust to fit vessels of different shapes and sizes.
The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Certifications: CE marked.
Ordering information: Three versatile platform options has to be ordered separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (76200-268) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty