Would you like to log in again?
SterEM™ SabDex (Sabouraud Dextrose) Agar, Irradiated, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W565
76200-288PK 136.54 USD InStock
76200-288
SterEM™ SabDex (Sabouraud Dextrose) Agar, Irradiated, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Sterile plates for environmental monitoring.
  • New moisture control packaging format
  • Prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping.
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation, validated for Sterility Assurance Level (SAL) of 10-5
  • Triple bagged two nylon inner bags and one outer Mylar® bag that is heat sealed
Sabouraud Dextrose Agar is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts, and meet the harmonized USP/EP/JP standards for the microbial examination of non sterile products. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Certified to ISO 13485. Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty