Would you like to log in again?
SterEM™ Tryptic Soy Agar (TSA), USP, with Lecithin and Tween® 80, Hardy Diagnostics
Supplier: Hardy Diagnostics
W520
76200-292PK 75.3 USD InStock
76200-292
SterEM™ Tryptic Soy Agar (TSA), USP, with Lecithin and Tween® 80, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Sterile plates for environmental monitoring.
  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation - Validated for Sterility Assurance Level (SAL) of 10-5
  • Triple bagged, two nylon inner bags and one outer Mylar(R) bag that is heat sealed
Tryptic Soy Agar (TSA) with Lecithin and Tween®, USP is recommended for use as a general growth medium for establishing microbiological trends, alerts and action levels in biologically controlled environments. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Certified to ISO 13485 - Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.
Ordering information: Minimum order of 16 packs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty