Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Kinesis® TELOS® QuEChERS Cleanup Kits, Cole-Parmer®
Supplier: Cole Parmer
06479-05 06479-09 06479-06 06478-67 06478-65 06478-80 06478-66 06479-13 06478-77 06479-10 06478-78 06479-07 06479-04 06478-79 06479-11 06479-12 06479-14 06478-68
76200-906PK 332.46 USD InStock
76200-906 76200-916 76200-908 76200-638 76200-902 76200-626 76200-904 76200-628 76200-634 76200-898 76200-900 76200-636 76200-910 76200-630 76200-640 76200-896 76200-632 76200-642
Kinesis® TELOS® QuEChERS Cleanup Kits, Cole-Parmer®
Chromatography Sorbents SPE Sorbents
Optimized to perform more advanced clean up of the sample.
  • Remove pigments, acids, and fats using three different combinations of SPE sorbents
  • Anhydrous magnesium sulfate removes any excess water
Further clean up of samples is easy with TELOS® QuEChERS clean up kits. After initial sample prep with the extraction kit, a more advanced clean up is often required for the removal of interferences such as pigments, organic acids and fats. The removal of these interferences is carried out using a combination of three different SPE sorbents.
TELOS® PestiCarb: removes pigments, including chlorophyll and carotenoids
TELOS® PSA: removes organic acids
TELOS® C18: removes nonpolar interferences, including fats
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty