Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Kinesis® TELOS® QuEChERS Extraction Kits, Cole-Parmer®
Supplier: Cole Parmer
06478-89 06478-86 06478-95 06478-90 06478-82 06478-88 06478-83 06478-93 06478-97 06479-00 06478-96 06478-92 06478-85 06479-02 06478-94 06478-87 06479-01
76200-608PK 184.87 USD InStock
76200-608 76200-616 76200-604 76200-912 76200-618 76200-606 76200-914 76200-612 76200-600 76200-622 76200-614 76200-602 76200-624 76200-596 76200-610 76200-598 76200-620
Kinesis® TELOS® QuEChERS Extraction Kits, Cole-Parmer®
Chromatography Sorbents SPE Sorbents
Quickly and economically extract the required analytes into solution.
  • Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
  • Break up complex sample types with homogenizer balls
TELOS® QuEChERS extraction kits offer the first stage cleanup for many sample types. This initial crude extraction and partitioning step uses organic solvent and salt solution to extract the required analytes into solution. Select from extraction kits without a tube, kits with a 50 ml centrifuge tube, or kits with a tube and homogenizer beads. Beads aid in breaking up more complex sample types.
Certifications: Conform with documented AOAC 2007.01 and EN 15662 protocols.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty