Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GloMax® Navigator Microplate Luminometer, Promega
Supplier: Promega
GloMax®
GM2000 GM2010
76201-022EA 23494.83 USD InStock
76201-022 76201-020
GloMax® Navigator Microplate Luminometer, Promega
Microplate Readers
The GloMax® navigator is an easy-to-use microplate luminometer integrated with Promega chemistries for superior luminescence assay performance. It is also possible to interpret results using integrated data analysis software.
  • Developed and optimized with Promega cell and gene reporter assays
  • Highest instrument sensitivity to measure the lowest assay signals
  • Low cross-talk between wells gives you better, more usable results
  • Over 30 preloaded assay protocols
  • Plate reading luminometer for convenient detection of bioluminescence
  • Top-reading, head-on photon counting photomultiplier tube detector
GloMax® navigator provides superior sensitivity and dynamic range for both strong and weak bioluminescence experimental samples as well as seamless integration with promega bioluminescence assays, including gene reporter, cell-based and biochemical assays. The microplate luminometer features optional dual injectors to measure both glow- and flash-based luminescent reactions in 96-well plates.
The luminometer is operated by an integrated tablet PC, which provides quick and easy navigation through the control options. Exporting your results is simple with a variety of options, including export to your local data network, USB flash drive and cloud-based storage location.
*Height of the instrument will be 18" with tablet and PC mounted.
Delivery information: One-year parts and labor warranty included.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty