Would you like to log in again?
ADP-Glo™ Max Assay, Promega
Supplier: Promega
V7001 V7002
76201-452EA 4884.89 USD InStock
76201-452 PAV7001
ADP-Glo™ Max Assay, Promega
Assays Enzyme Assays
The ADP-Glo Max Assay is a luminescent ADP detection assay that provides a universal, homogeneous, high-throughput screening method to measure ATPase or kinase activity by quantifying the amount of ADP produced in a reaction.
  • Luminescent Method to Measure ATPase or Kinase Activity
  • Screen library compounds for effects on target enzymes
  • Perform mode of action studies on enzymes of interest
  • Measure activity of immunoprecipitated ATPases/kinases
  • Use when higher ATP concentration is required (up to 5mM)
The ADP-Glo Max Assay is a luminescent ADP detection assay that provides a universal, homogeneous, high-throughput screening method to measure ATPase or kinase activity by quantifying the amount of ADP produced in a reaction. The assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) when higher ATP concentration is required (up to 5mM). The ADP-Glo Max Assay produces a strong signal that positively correlates with enzyme activity and can be adapted to a multitude of plate formats. The assay is performed in two steps: first, after the completion of the ADP-producing reaction, an equal volume of ADP-Glo Reagent is added to terminate the reaction and deplete the remaining ATP. Second, the ADP-Glo Max Detection Reagent is added to simultaneously convert ADP to ATP, and the latter is converted to light in a coupled reaction with luciferase/luciferin. The ADP-Glo Max Assay has a high dynamic range and produces a strong signal at low ATP to ADP conversion, making it well suited for screening low-activity ATPases such as drug membrane transporters and heat shock proteins. The assay produces minimal false hits and Z' values of greater than 0.7. Several Kinase Enzyme Systems are available. Visit www.promega.com/kinase/ to see the collection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty