Would you like to log in again?
Glucose Uptake-Glo™ Assay, Promega
Supplier: Promega
J1341
76201-554EA 517.34 USD InStock
76201-554
Glucose Uptake-Glo™ Assay, Promega
Assays Cellular Assays
A simple, non-radioactive assay used to measure glucose uptake inside cells.
  • Homogeneous protocol
  • Achieve sensitivity with broad linearity
  • Reliable and reproducible results
Glucose metabolism is a key process in many organisms. A lack of insulin-stimulated glucose uptake is associated with type 2 diabetes, while high glucose uptake is a sign of the high glycolytic rates associated with cancer. Measuring glucose uptake can determine the effects of various treatments for diabetes and cancer.
The gold standard method of assaying glucose uptake relies on detection of radio-labelled glucose analogs. Alternative colorimetric and fluorometric glucose uptake assays often lack the sensitivity and robustness needed to reliably measure glucose uptake in cells.
Delivery information: The system contains sufficient reagents to perform 50 reactions in 96-well plates (50 µl of sample, 25 µl of stop buffer, 25 µl of neutralization buffer, 100 µl of 2DG6P detection reagent). It includes luciferase reagent (5 ml), stop buffer (15 ml), neutralization buffer (15 ml), NADP+ (20mM) (50 μl), glucose-6-phosphate dehydrogenase (G6PDH) (125 μl), reductase (25 μl), reductase substrate (55 μl), 2-deoxyglucose (2DG, 100mM) (250 μl), 2DG6P standard (1mM) (50 μl). Store the kit at –65 °C.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty