Would you like to log in again?
Nano-Glo® Live Cell Assay System, Promega
Supplier: Promega
N2012 N2013 N2011
76201-574EA 12067.5 USD InStock
76201-574 76201-572 76201-570
Nano-Glo® Live Cell Assay System, Promega
Assays Cellular Assays
The Nano-Glo® Live Cell Assay System is a single-addition, non-lytic detection reagent used to measure NanoBiT® or NanoLuc® luminescence from living cells.
  • Substrate and buffer optimized to enhance stability
  • Use to detect NanoBiT® protein complementation or NanoLuc® reporter activity
  • Monitors luminescence at a single time point or continuously for up to 2 hours without compromising cell viability
The reagent is prepared by diluting the Nano-Glo® Live Cell Substrate with the Nano-Glo® LCS Dilution Buffer to make the Nano-Glo® Live Cell Reagent, a 5X stock that is added directly to cell culture medium. Both substrate and buffer solutions are optimized to provide enhanced stability.
The Nano-Glo® Live Cell Reagent is designed to reduce autoluminescence in the presence or absence of serum, increasing the sensitivity for detection of low levels of NanoBiT® or NanoLuc® luminescence. The Nano-Glo® Live Cell Assay System can be used to monitor luminescence at a user-defined time point or continuously for up to 2 hours without compromising cell viability.
The signal stability observed after addition of the Nano-Glo® Live Cell Reagent to culture media can be influenced by experimental parameters such as high levels of enzyme activity, temperature or the presence of FBS in the media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty