Would you like to log in again?
GLOVE LIFESTAR EC LSE-104 NTRL M CS1000
Supplier: Ansell Healthcare
Lifestar Ec* Exam Glove Medium^ Lifestar Ec* Exam Glove, Material: Nitrile, Color: Blue Interior / White Exterior, Powder Mass: Max. 2.0 Mg, Double Dipped, Extended Cuff, Textured Fingers, Meets Nfpa Standards, Exceptional Comfort & Tactility, Non-Stick Formulation, Length: 12 Inch, Size: Medium
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE LIFESTAR EC LSE-104 NTRL M CS1000 76201-714 LSE-104-M Box of 100 Retrieving
Case of 1000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty