Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RNA polymerase II Mouse Monoclonal Antibody [clone: 8WG16]

Supplier: Biosensis
ABS132A-0100 C-1655-100
76201-798EA 579 USD InStock
76201-798 76264-602
Anti-RNA polymerase II Mouse Monoclonal Antibody [clone: 8WG16]
Antibodies
Recommended Applications: IF, IP, WB

Species reactivity: Species independent

Type: Primary
Antigen: RNA polymerase II
Clonality: Monoclonal
Clone: 8WG16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty