Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EpCAM Mouse Monoclonal Antibody [clone: rMOC-31]

Supplier: Enzo Life Sciences
ABS317-0100
76201-876EA 545 USD InStock
76201-876
Anti-EpCAM Mouse Monoclonal Antibody [clone: rMOC-31]
Antibodies
Recombinant monoclonal antibody

  • Recommended Applications: Flow Cytometry, IF, IHC (PS), WB
  • Species reactivity: Human

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: rMOC-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: lgG1κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty