Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD44 Rabbit Monoclonal Antibody [clone: CD44v4/1700R]

Supplier: Enzo Life Sciences
ABS349-0100
76201-946EA 677 USD InStock
76201-946
Anti-CD44 Rabbit Monoclonal Antibody [clone: CD44v4/1700R]
Antibodies
Recombinant monoclonal antibody

  • Recommended Applications: Flow Cytometry, IF, IHC (PS)
  • Species reactivity: Human

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: CD44v4/1700R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty