Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (human female genomic)

Supplier: Enzo Life Sciences

GEN118-0100
76202-256EA 225 USD InStock
76202-256
DNA (Deoxyribonucleic acid) (human female genomic)
DNA (Deoxyribonucleic acid)
A normal pool of female genomic DNA that can serve as a standard in molecular biology applications

  • Concentration: 150 - 250 µg/mL in 10 mM Tris-HCl, pH 8.0, containing 1 mM EDTA

Human Genomic DNA, Female, is comprised of a pool of ≥ 8 normal donors. It is intact DNA extracted from freshly harvested whole blood of healthy females. DNA is treated with DNase free-RNase to remove residual contaminant RNA.

Human genomic DNA can be used for a variety of molecular applications including PCR, southern blotting, and DNA library construction. It can also be used as normal reference DNA for comparative genomic hybridizations (CGH).


MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty