Would you like to log in again?
Arthrobacter luteus Zymolyase
Supplier: MP Biomedicals
ImmunO™
08320932 08320931
IC320932EA 1437.16 USD InStock
IC320932 76202-274
Arthrobacter luteus Zymolyase
Enzymes
Zymolyase-100T is an enzyme preparation from a submerged culture of Arthrobacter luteus which effectively lyses cell walls of viable yeast cells.
  • Lyophilized enzyme partially purified by affinity chromatography
  • Contains 100,000 units enzyme/g
  • Primary enzyme responsible for lysis of viable yeast cells in ß-1,3-glucan laminaripentaohydrolase
  • Hydrolyzes glucose polymers at the ß-1,3-glucan linkages releasing laminaripentaose as the principal product
Zymolyase 100T is used to lyse living yeast cell walls to produce protoplast or spheroplast of various strains of yeast cells.
The lytic spectrum of this enzyme includes organisms from the following genera: Ashbya, Candida, Debaryomyces, Eremothecium, Endomyces, Hansenula, Hanseniaspora, Kloekera, Kluyveromyces, Lipomyces, Metschikowia, Pichia, Pullularia, Torulopsis, Saccharomyces, Saccharomcopsis, Saccharomycodes, Schwannimomyces and others.
Activators: Dithiothreitol, 2-mercaptoethanol and other sulfhydryl compounds (such as cysteine); Essential Enzyme: ß-1,3-glucan laminaripentaohydrolase; Other Enzymes: ß-1,3-glucanase, ca. 1.5×107 units/g, Protease, ca. 1.0×104 units/g, Mannase, ca. 1.0×104 units/g.
Zymolase-100T has a low solubility. Use as a suspension. If it is necessary to make a sterile enzyme solution of more than 0.05%, make a 2% enzyme stock solution by dissolving Zymolase -100T in a buffer solution (pH 7.5) which contains 5% glucose. To remove insoluble substances, filter through a sterile filter. Do not use Nitrocellulose filters. Dilute sterile enzyme suspension to desired concentration.
It may also be used for glucan hydrolysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty