Would you like to log in again?
BIN STACK/HANG PLASTIC BLUE 5X4-1/8X3 IN
Supplier: QUANTUM STORAGE SYSTEMS
Ultra stack and Hang bin 5 x 4-1/8 x 3in^ Ultra stack and Hang bin, Material: Made from strong and durable injection molded Plastic, Color: Blue, Autoclavable up to 250 deg F, as well as being resistant to extreme cold, 1 Divider slot, ID: 4-3/8x3-7/16x2-13/16in, Outer Dimension: 5 x 4-1/8 x 3in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STACK/HANG PLASTIC BLUE 5X4-1/8X3 IN 76202-694 QUS200BL Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty