Would you like to log in again?
BIN STACK/HANG PP IVORY 10-7/8X4-1/8X4IN
Supplier: QUANTUM STORAGE SYSTEMS
Ultra stack and Hang bin 10-7/8 x 4-1/8 x 4in^ Ultra stack and Hang bin, Material: Injection molded polypropylene, Color: Ivory, Has molded in back and side grips, Resistant to extreme cold, 1 Divider slot, Inner Dimension: 10-1/4 x 3-3/16 x 3-3/4in, Outer Dimension: 10-7/8 x 4-1/8 x 4in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STACK/HANG PP IVORY 10-7/8X4-1/8X4IN 76202-732 QUS224IV Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty