Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BIN STACKHANG PP BLACK 7-3/8X4-1/8X3IN
Supplier: QUANTUM STORAGE SYSTEMS
Ultra stack and Hang bin 7-3/8 x 4-1/8 x 3in^ Ultra stack and Hang bin, Material: Injection molded polypropylene, Color: Black, Back and side grips make it easy to transport and move, 1 Divider slot, Resistant to extreme cold, ID: 6-3/4x 3-7/16 x 2-13/16in, Outer Dimension: 7-3/8 x 4-1/8 x 3in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STACKHANG PP BLACK 7-3/8X4-1/8X3IN 76202-798 QUS220BK Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty