Would you like to log in again?
BIN STACK HANG PP GREEN 16 X11 X 8 IN
Supplier: QUANTUM STORAGE SYSTEMS
Ultra stack and Hang bin 16 x 11 x 8in^ Ultra stack and Hang bin, Material: Injection molded polypropylen, Color: Green, Open hopper front, Has molded in back and side grips, 3 Divider slots, Resistant to extreme cold, Inner Dimension: 15-1/8 x 10 x 7-3/4in, Outer Dimension: 16 x 11 x 8in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STACK HANG PP GREEN 16 X11 X 8 IN 76202-864 QUS255GN Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty