Would you like to log in again?
BIN STACK HANG PP GREEN 9 1/4 X6 X5 IN
Supplier: QUANTUM STORAGE SYSTEMS
Ultra stack and Hang bin 9-1/4 x 6 x 5in^ Ultra stack and Hang bin, Material: Injection molded polypropylene, Color: Green, Tough and durable, Back and side grips make it easy to move around, 1 Divider Slot, Resistant to extreme cold, ID: 8-1/2x 5-1/8x 4-1/2in, Outer Dimension: 9-1/4 x 6 x 5in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIN STACK HANG PP GREEN 9 1/4 X6 X5 IN 76202-908 QUS221GN Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty