Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Grade 934-AH™ RTU Glass Microfiber Filters, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

934-AH™ Whatman™ 934-AH™
4827-070 9927-047 2827-047 9907-042 9907-090 9927-090 2827-047 4827-035 4827-070 3827-090 3827-047 9927-070 3827-042 4827-035 3827-042 9907-055 4827-042 9907-047 4827-090 9907-047 9927-047 3827-070 9907-070 9927-070 9907-9436 4827-042 9927-090 3827-035 9907-9436 4827-047 4827-090 3827-090 3827-070 3827-035 9907-070 9907-090 9907-055 9907-042 4827-047 3827-047
76202-944PK 109.81 USD InStock
CA97040-978 CA97040-974 CA76203-004 76202-944 CA76202-996 CA76203-002 76202-946 CA76202-998 CA76203-000 76202-948 75874-050 CA11025-656 CA11008-600 76202-950 CA76202-984 76202-930 76202-952 89410-170 76097-936 97040-978 97040-976 CA76202-986 76202-954 97040-974 76202-932 CA76202-988 76202-956 CA75874-050 76202-934 76202-936 CA76203-010 76202-958 CA76202-990 CA76203-008 76202-940 CA76202-992 CA76203-006 76202-942 CA76202-994 CA76097-936
Whatman™ Grade 934-AH™ RTU Glass Microfiber Filters, Whatman products (Cytiva)
Filters Glass Fiber Filters
These 934-AH™ RTU grade filter papers have been pre-washed, dried (or ignited), cooled, desiccated, and weighed. This eliminates the pre-treating steps that are otherwise required by the US EPA Laboratory Standard Method 2540 parts C, D, and E.

  • Superior fine particle retention at high flow rates
  • Capable of processing very turbid samples
  • Particle retention: 1.5 µm

These filter papers are widely used for dissolved, suspended and volatiles solid analysis. They are also used as a wastewater filter for a wide range of water monitoring applications, including the monitoring of rivers, lakes, and coastal waters, as well as analyzing purified discharge of wastewater treatment plants and monitoring discharge water from industrial plants.

934-AH™ RTU for Suspended and Dissolved Solids: Pre-washed, dried, cooled, and weighed filters with certified mass loss between washing and drying steps of the lesser of less than 0.5 mg or 4% in accordance with US EPA Lab Standard Method 2540 parts C and D. Most filters come weighed to four decimal places, however a version of the 47 mm filter can be measured to five decimal places if needed.

934-AH™ RTU for Volatiles: Pre-washed, pre-weighed, and pre-fired at 550 °C aligned to US EPA Laboratory Standard Method 2540 parts C, D, and E.

934-AH™ RTU Double Weigh: Drying, cooling, and weighing steps repeated twice for customers with the highest regulatory concern.

Packaging: The pre-weighed filter papers are packaged within an aluminium pan with the filter weight clearly printed on a heat resistant label. Each pan has a unique barcode. Each box also has a barcode that can be used to download the weights of all filters from that box.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty