Would you like to log in again?
Whatman™ Grade GF/C RTU Glass Microfibre Filters without Binder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
2822-047 3822-090 3822-070 2822-090 2822-070 3822-090 3822-047 2822-070 3822-047 2822-090 2822-047 3822-070
76202-960PK 119.57 USD InStock
76202-960 CA76202-980 76202-970 CA76202-982 76202-962 76202-964 CA76202-972 76202-966 CA76202-974 76202-968 CA76202-976 CA76202-978
Whatman™ Grade GF/C RTU Glass Microfibre Filters without Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water.
  • Fine particle retention
  • Good flow rate and loading capacity
  • Binder free
  • Particle retention: 1.2 µm
The GF/C RTU line is the pre-treated version of the traditional GF/C filters. They are washed, dryed, cooled (in a desiccator), and weighted. These filters are certified to have weight loss of less than 0.017 mg/cm² after preparation, which is required by the standard. Each ready-to-use filter is placed in an individually barcoded aluminium pan that notes the filter weight. The barcode can be scanned to give customers more information regarding the weight of the filter.
Pre-rinsed, dried and un-weighed versions are also available.
Delivery information: Each box contains an additional box ID and barcode that can be used to download the weights of all 100 pans in that box.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty