Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LDK378 ≥99% (by HPLC)

Supplier: Enzo Life Sciences
Synonyms: LDK-3785-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamineCeritinib (free base)5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulphonyl)phenyl)pyrimidine-2,4-diamine

CHM218-0025 CHM218-0500 CHM218-0050 CHM218-0005 CHM218-0200 CHM218-0100 CHM218-0010
76203-130EA 186.25 USD InStock
76203-130 76203-140 76203-128 76203-138 76203-136 76203-134 76203-132
LDK378 ≥99% (by HPLC)
LDK378
Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer.

  • Selective ALK inhibitor
  • Off-white solid
  • Soluble in DMSO.


Formula: C₂₈H₃₆ClN₅O₃S
MW: 558.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26142648
CAS Number: 1032900-25-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/17/05/45401705.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty