Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Speed Crank Adjustable Stand Up Desks, 60", Luxor

Supplier: Luxor
STANDCF60-BKBO STANDCF60-AGBO
76203-774EA 1350.34 USD InStock
76203-774 76203-772
High Speed Crank Adjustable Stand Up Desks, 60", Luxor
Furniture Tables and Workbenches
Quickly deliver collaborative and productive comfort to your workday by seamlessly alternating between sitting and standing with the Luxor pneumatic adjustable height standing desk.

  • High speed crank allows for rapid adjustment from sitting to standing
  • Crank handle quickly raises the desk height between 32.6" and 48.5" (with casters)
  • Crank handle quickly raises the desk height between 29.6" and 45.5" (with glides)
  • Spacious 59×29.5" desktop provides plenty of room for multiple monitor setups
  • Includes choice of leveling glides and 3" casters, two with locking brake
  • Max weight capacity: 154 lbs.

Now available with a high-speed crank operated lift mechanism, the turn-handle crank quickly moves the desk from the lowest to highest positions within seconds without requiring electric power.

The generous 59×29.5" work surface easily fits triple monitors plus a full-sized keyboard and mouse while the height adjustment range accommodates taller and shorter users. Each desk includes a choice of leveling glides as well as 3" casters, two with locking brakes, allowing for easy maneuverability to suit ever-changing workspace needs.

Enjoy the health benefits of standing while increasing focus and productivity with a high-speed crank adjustable desk from Luxor.

Overall with casters: W59×D29.5×H32.6 - 48.5";
Overall with glides: W59×D29.5×H29.6 - 45.5";
Desktop: W59×D29.5"

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty