Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adjustable Flip-Top Table, Crank Handle, 60", Luxor

Supplier: Luxor
STAND-NESTC-60
76203-784EA 1425.21 USD InStock
76203-784
Adjustable Flip-Top Table, Crank Handle, 60", Luxor
Furniture Tables and Workbenches
The Luxor flip-top table is a multipurpose sit or stand desk that's perfect for schools, training rooms or general office use.

  • Crank handle offers an economical choice without compromising quality
  • Adjusts between 30" and 45.25" for seated and standing users
  • Table is lightweight and moves easily on four 3" furniture casters (two locking brakes)
  • Easy setup and teardown with release latches under table
  • Weight capacity of 150 lbs.
  • Grommet holes route, network and organize cables

The convenient crank handle adjusts the table height from 30" to 42.25" to accommodate a range of seated or standing users, while 3" locking casters allow for easy mobility. Adding to its versatility, each table can be neatly folded for space-saving storage using two pull handles that flip the table into a nesting position. Choose the Luxor 60" Adjustable Flip Top Table to create a professional, ergonomic workspace.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty