Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wall-Mounted Magnetic Whiteboard , 48W×48" H, Luxor

Supplier: Luxor
WB4848W
76203-814EA 336.87 USD InStock
76203-814
Wall-Mounted Magnetic Whiteboard , 48W×48" H, Luxor
Note Boards
Built for brainstorming, teaching or organizing ideas and information, a Luxor magnetic dry erase board doubles the writing space needed for dynamic workspaces.

  • Economically priced without compromising quality
  • Painted steel surface resists ghosting and provides excellent erasability
  • Aluminum frame(1") gives board a durable, sleek look
  • Magnetic surface operates as a bulletin board for additional communication
  • Deep aluminum marker/eraser tray(2") provides ample storage
  • Includes mounting brackets and hardware

More than a surface for brainstorming, teaching and illustrating ideas, wall-mounted dry erase boards become a focal point of any room. That's why Luxor wall-mounted magnetic whiteboards offer superior performance in a modern style at an affordable price. This square whiteboard has a high-quality, ghosting-resistant painted steel surface for bright visibility, high durability, and can also be used as a magnetic bulletin board. The slim aluminum frame and marker/eraser tray offer lightweight durability designed to last for use after use, year after year.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty