Would you like to log in again?
CRITERION™ Shigella Broth Base, Dehydrated Culture Media
Supplier: Hardy Diagnostics
CRITERION™
C9151 C9151
76203-716EA 141.34 USD InStock
76203-716 76203-908
CRITERION™ Shigella Broth Base, Dehydrated Culture Media
Media for Microbiology Base Materials for Microbiology
This media is recommended for use in the selective enrichment and cultivation of food samples.
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth
Delivery information: Container sizes include 2 L mylar® bag – pre-measured for a 2 L batch, no weighing needed, 500 g bottle - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.
Caution: Not intended to be used for the diagnosis of human disease.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty